Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001166391.1 | NP_001159863.1 | 8 | 1076 | |
NM_028784.3 | NP_083060.2 | 8 | 1101 | ||
GenBank mRNA | AK030914.1 | - | 8 | 1076 | |
AK036403.1 | - | 8 | 1102 | ||
AK049092.1 | - | 8 | 1102 | ||
AK163591.1 | - | 8 | 1076 | ||
AK165311.1 | - | 8 | 1102 | ||
BC011073.1 | - | 2 | 182 | ||
BC040274.1 | - | 8 | 1092 | 3482233 |