Gene Transcripts
- Gene Symbol
- Ppp1r2
- Entrez Gene ID
- 66849
- Gene Name
- protein phosphatase 1, regulatory (inhibitor) subunit 2
- Gene Aliases
- 0610025N14Rik, 2310007G06Rik, 4930440J04Rik, 5430408E15Rik, D16Ertd248e, IPP-2
- Chromosome Location
- Chr. 16: 31251541 - 31275277 on Build GRCm38
- UniGene ID
- Mm.291593
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_025800.3 |
NP_080076.1
|
5, 6 |
889 |
|
GenBank mRNA |
AK002671.1 |
- |
5, 6 |
836 |
|
AK004320.1 |
- |
4, 5 |
459 |
|
AK015349.1 |
- |
5, 6 |
717 |
|
AK015709.1 |
- |
|
666 |
|
AK032462.1 |
- |
5, 6 |
840 |
|
AK053657.1 |
- |
5, 6 |
790 |
|
AK077194.1 |
- |
5, 6 |
714 |
|
AK150281.1 |
- |
5, 6 |
892 |
|
BC069885.1 |
- |
5, 6 |
862 |
3671082 |
BC069886.1 |
- |
5, 6 |
834 |
6310133 |
- Gene Symbol
- Ppp1r2-ps3
- Entrez Gene ID
- 546723
- Gene Name
- protein phosphatase 1, regulatory (inhibitor) subunit 2, pseudogene 3
- Gene Aliases
- EG546723, Gm5972
- Chromosome Location
- Chr. 19: 30538876 - 30539679 on Build GRCm38
- UniGene ID
- Mm.301509
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC100445.1 |
- |
1 |
713 |
|
View Full Product Details