Gene Transcripts

Gene Symbol
Hmg20a
Entrez Gene ID
66867
Gene Name
high mobility group 20A
Gene Aliases
1200004E06Rik, 5730490E10Rik, Hmgxb1, Ibraf
Chromosome Location
Chr. 9: 56418628 - 56496936 on Build GRCm38
UniGene ID
Mm.150856
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025812.2 NP_080088.1 4 373
GenBank mRNA AK004596.1 - 4 393
AK017716.1 - 4 576
AK032790.1 - 4 401
AK039222.1 - 4 395
AK129010.1 - 4 575
AK134213.1 - 4 545
AL355705.1 - 4 359
BC013804.1 - 4 373 3602239
BC068257.1 - 4 446 6434284
DQ182735.1 - 4 373

View Full Product Details