Gene Transcripts

Gene Symbol
Fadd
Entrez Gene ID
266610
Gene Name
Fas (TNFRSF6)-associated via death domain
Gene Aliases
Mort1
Chromosome Location
Chr. 1: 217746176 - 217748581 on Build Rnor_6.0
UniGene ID
Rn.16183
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152937.2 NP_690920.1 2 584
GenBank mRNA AF406779.1 - 2 584
AJ441127.1 - 609

View Full Product Details