Gene Transcripts

Gene Symbol
RAC1
Entrez Gene ID
5879
Gene Name
ras-related C3 botulinum toxin substrate 1 (rho family, small GTP binding protein Rac1)
Gene Aliases
MIG5, Rac-1, TC-25, p21-Rac1
Chromosome Location
Chr. 7: 6374495 - 6403967 on Build GRCh38
UniGene ID
Hs.413812
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006908.4 NP_008839.2 6 1342
NM_018890.3 NP_061485.1 7 1399
GenBank mRNA AK054993.1 - 6 1317
BC050687.1 - 6 1289 6149377
D25274.1 - 1 233

View Full Product Details