Gene Transcripts

Gene Symbol
SFRP2
Entrez Gene ID
6423
Gene Name
secreted frizzled related protein 2
Gene Aliases
FRP-2, SARP1, SDF-5
Chromosome Location
Chr. 4: 153780590 - 153789076 on Build GRCh38
UniGene ID
Hs.481022
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003013.2 NP_003004.1 3 1430
GenBank mRNA AF311912.1 - 3 1426
AK075372.1 - 3 1423
AK093922.1 - 4 1398

View Full Product Details