Gene Transcripts
- Gene Symbol
- COQ2
- Entrez Gene ID
- 27235
- Gene Name
- coenzyme Q2, polyprenyltransferase
- Gene Aliases
- CL640, COQ10D1, MSA1, PHB:PPT
- Chromosome Location
- Chr. 4: 83138568 - 83285129 on Build GRCh38
- UniGene ID
- Hs.144304
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_015697.7 |
NP_056512.5
|
5, 6 |
920 |
|
XM_011531855.2 |
XP_011530157.1
|
5, 6 |
1131 |
|
XM_011531857.2 |
XP_011530159.1
|
5, 6 |
1129 |
|
XM_011531859.2 |
XP_011530161.1
|
5, 6 |
1128 |
|
XM_011531860.2 |
XP_011530162.1
|
5, 6 |
1128 |
|
XM_011531862.2 |
XP_011530164.1
|
5, 6 |
1127 |
|
XM_011531863.2 |
XP_011530165.1
|
5, 6 |
1124 |
|
XM_011531866.2 |
XP_011530168.1
|
5, 6 |
1120 |
|
XM_011531867.2 |
XP_011530169.1
|
5, 6 |
1043 |
|
XM_017008031.1 |
XP_016863520.1
|
5, 6 |
1043 |
|
XM_017008032.1 |
XP_016863521.1
|
3, 4 |
313 |
|
GenBank mRNA |
AF091086.1 |
- |
4, 5 |
469 |
|
AK129583.1 |
- |
5, 6 |
676 |
|
BC008804.2 |
- |
5, 6 |
777 |
3954787 |
BC020728.1 |
- |
5, 6 |
817 |
4696019 |
BQ363759.1 |
- |
2, 3 |
196 |
|
CR456860.1 |
- |
2, 3 |
179 |
|
View Full Product Details