Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001308165.1 | NP_001295094.1 | 3 | 367 | |
NM_007017.2 | NP_008948.1 | 2 | 1470 | ||
NM_178424.1 | NP_848511.1 | 2 | 1470 | ||
XM_005265803.4 | XP_005265860.1 | 3 | 234 | ||
XM_011534420.1 | XP_011532722.1 | 3 | 1260 | ||
GenBank mRNA | AB022083.1 | - | 2 | 1423 | |
AB022441.1 | - | 2 | 1423 | ||
AK302162.1 | - | 3 | 367 | ||
AK308329.1 | - | 2 | 1418 | ||
BC033492.1 | - | 2 | 1470 | 5172577 |