Gene Transcripts
- Gene Symbol
- CEL
- Entrez Gene ID
- 1056
- Gene Name
- carboxyl ester lipase
- Gene Aliases
- BAL, BSDL, BSSL, CELL, CEase, FAP, FAPP, LIPA, MODY8
- Chromosome Location
- Chr. 9: 133061978 - 133071863 on Build GRCh38
- UniGene ID
- Hs.533258
- Species
- Homo sapiens
- Gene Symbol
- CELP
- Entrez Gene ID
- 1057
- Gene Name
- carboxyl ester lipase pseudogene
- Gene Aliases
- CELL, cell1, cell2, cell3
- Chromosome Location
- Chr. 9: 133082539 - 133087091 on Build GRCh38
- UniGene ID
- Hs.654361
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_001275.2 |
|
2 |
133 |
|
GenBank mRNA |
L14813.1 |
- |
2 |
133 |
|
View Full Product Details