Gene Transcripts
- Gene Symbol
- GPAT2
- Entrez Gene ID
- 150763
- Gene Name
- glycerol-3-phosphate acyltransferase 2, mitochondrial
- Gene Aliases
- CT123
- Chromosome Location
- Chr. 2: 96021946 - 96035974 on Build GRCh38
- UniGene ID
- Hs.348629
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001321525.1 |
NP_001308454.1
|
3, 4 |
587 |
|
NM_001321526.1 |
NP_001308455.1
|
3, 4 |
677 |
|
NM_001321527.1 |
NP_001308456.1
|
3, 4 |
641 |
|
NM_001321528.1 |
NP_001308457.1
|
3, 4 |
641 |
|
NM_001321529.1 |
NP_001308458.1
|
3, 4 |
677 |
|
NM_001321530.1 |
NP_001308459.1
|
3, 4 |
587 |
|
NM_001321531.1 |
NP_001308460.1
|
4, 5 |
648 |
|
NM_207328.3 |
NP_997211.2
|
3, 4 |
677 |
|
XM_005263888.3 |
XP_005263945.1
|
3, 4 |
352 |
|
XM_006712308.3 |
XP_006712371.1
|
3, 4 |
352 |
|
XM_006712311.3 |
XP_006712374.1
|
3, 4 |
352 |
|
XM_006712312.3 |
XP_006712375.1
|
3, 4 |
352 |
|
XM_006712313.3 |
XP_006712376.1
|
3, 4 |
352 |
|
XM_017003424.1 |
XP_016858913.1
|
3, 4 |
291 |
|
XM_017003425.1 |
XP_016858914.1
|
3, 4 |
352 |
|
XM_017003426.1 |
XP_016858915.1
|
3, 4 |
352 |
|
GenBank mRNA |
AK131194.1 |
- |
4, 5 |
648 |
|
AK298131.1 |
- |
3, 4 |
267 |
|
AK302887.1 |
- |
3, 4 |
312 |
|
AK302918.1 |
- |
3, 4 |
323 |
|
BC068596.1 |
- |
3, 4 |
348 |
4837765 |
CB961934.1 |
- |
3, 4 |
339 |
|
View Full Product Details