Gene Transcripts
- Gene Symbol
- CTCFL
- Entrez Gene ID
- 140690
- Gene Name
- CCCTC-binding factor like
- Gene Aliases
- BORIS, CT27, CTCF-T, HMGB1L1, dJ579F20.2
- Chromosome Location
- Chr. 20: 57495965 - 57525652 on Build GRCh38
- UniGene ID
- Hs.131543
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC137482.1 |
- |
1 |
367 |
|
BC137483.1 |
- |
1 |
367 |
9021483 |
View Full Product Details