Gene Transcripts
- Gene Symbol
- KANSL2
- Entrez Gene ID
- 54934
- Gene Name
- KAT8 regulatory NSL complex subunit 2
- Gene Aliases
- C12orf41, NSL2
- Chromosome Location
- Chr. 12: 48653211 - 48682252 on Build GRCh38
- UniGene ID
- Hs.505412
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK000443.1 |
- |
7, 8 |
979 |
|
View Full Product Details