Gene Transcripts

Gene Symbol
PPP1R27
Entrez Gene ID
116729
Gene Name
protein phosphatase 1 regulatory subunit 27
Gene Aliases
DYSFIP1
Chromosome Location
Chr. 17: 81833492 - 81835050 on Build GRCh38
UniGene ID
Hs.454720
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007533.3 NP_001007534.1 3 755
GenBank mRNA AF434846.1 - 3 733
AK096888.1 - 2 1405
BI826571.1 - 2 376

View Full Product Details