Gene Transcripts

Gene Symbol
Tspyl2
Entrez Gene ID
52808
Gene Name
TSPY-like 2
Gene Aliases
CDA1, CINAP, DENTT, DXBwg1396e, DXHXS1008E, E130307F10Rik
Chromosome Location
Chr. X: 152336852 - 152342506 on Build GRCm38
UniGene ID
Mm.9997
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029836.3 NP_084112.1 6 1753
GenBank mRNA BC030334.1 - 4 900
BC043694.1 - 6 1467
BC054393.1 - 6 1673 6491672

View Full Product Details