Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001099666.1 | NP_001093136.1 | 5 | 608 | |
XM_005251976.4 | XP_005252033.1 | 5 | 698 | ||
XM_005251977.4 | XP_005252034.1 | 5 | 672 | ||
XM_005251980.4 | XP_005252037.1 | 5 | 658 | ||
XM_005251981.4 | XP_005252038.1 | 5 | 658 | ||
XM_011518639.1 | XP_011516941.1 | 5 | 572 | ||
XM_011518640.1 | XP_011516942.1 | 6 | 748 | ||
GenBank mRNA | AL832683.1 | - | 3 | 352 | |
BC053622.1 | - | 5 | 590 |