Gene Transcripts

Gene Symbol
PDSS2
Entrez Gene ID
57107
Gene Name
prenyl (decaprenyl) diphosphate synthase, subunit 2
Gene Aliases
C6orf210, COQ10D3, DLP1, bA59I9.3, hDLP1
Chromosome Location
Chr. 6: 107152557 - 107459683 on Build GRCh38
UniGene ID
Hs.745008
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020381.3 NP_065114.3 8 1783
XM_011535956.2 XP_011534258.1 9 2079
XM_011535957.2 XP_011534259.1 8 1957
XM_011535958.2 XP_011534260.1 8 1945
XM_011535959.2 XP_011534261.1 7 1724
XM_011535960.2 XP_011534262.1 9 1708
XM_011535961.2 XP_011534263.1 6 1497
XM_011535962.2 XP_011534264.1 8 1425
GenBank mRNA AF254956.1 - 8 1754
AL832290.1 - 3 588
BC039906.1 - 8 1769 6066481

View Full Product Details