Gene Transcripts

Gene Symbol
Olr865
Entrez Gene ID
298474
Gene Name
olfactory receptor 865
Gene Aliases
Chromosome Location
Chr. 5: 137812062 - 137815850 on Build Rnor_6.0
UniGene ID
Rn.143175
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000410.1 NP_001000410.1 1 343
XM_008763996.1 XP_008762218.1 1 343

View Full Product Details