Gene Transcripts

Gene Symbol
DGAT2
Entrez Gene ID
84649
Gene Name
diacylglycerol O-acyltransferase 2
Gene Aliases
ARAT, GS1999FULL, HMFN1045
Chromosome Location
Chr. 11: 75768733 - 75801536 on Build GRCh38
UniGene ID
Hs.129798
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001253891.1 NP_001240820.1 4 599
NM_032564.4 NP_115953.2 5 728
XM_011545304.2 XP_011543606.1 5 432
GenBank mRNA AB048286.2 - 5 699
AB073384.1 - 4 276
AF384161.2 - 5 476
AK091870.1 - 1 1231
AL834287.1 - 5 607
AY358532.1 - 5 653
AY780647.1 - 4 331
BC015234.1 - 5 717 3903313
CR749377.1 - 1 4212

View Full Product Details