Gene Transcripts
- Gene Symbol
- ENSA
- Entrez Gene ID
- 2029
- Gene Name
- endosulfine alpha
- Gene Aliases
- ARPP-19e
- Chromosome Location
- Chr. 1: 150622123 - 150629622 on Build GRCh38
- UniGene ID
- Hs.632456
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_004436.2 |
NP_004427.1
|
2, 3 |
328 |
|
NM_207044.1 |
NP_996927.1
|
2, 3 |
328 |
|
NM_207046.1 |
NP_996929.1
|
2, 3 |
192 |
|
NM_207047.1 |
NP_996930.1
|
2, 3 |
192 |
|
GenBank mRNA |
AF067170.1 |
- |
2, 3 |
285 |
|
AF157509.1 |
- |
2, 3 |
282 |
|
AF157510.1 |
- |
2, 3 |
282 |
|
AK001981.1 |
- |
2, 3 |
302 |
|
AK290064.1 |
- |
2, 3 |
175 |
|
AL711949.1 |
- |
2, 3 |
331 |
|
BC000436.2 |
- |
2, 3 |
276 |
2820528 |
BC004461.1 |
- |
2, 3 |
276 |
2820528 |
BC068544.1 |
- |
2, 3 |
293 |
5273162 |
BE799496.1 |
- |
2, 3 |
212 |
|
BQ883543.1 |
- |
2, 3 |
290 |
|
CR457130.1 |
- |
2, 3 |
176 |
|
CR536578.1 |
- |
2, 3 |
176 |
|
T78564.1 |
- |
2, 3 |
208 |
|
X99906.1 |
- |
2, 3 |
301 |
|
View Full Product Details