Gene Transcripts

Gene Symbol
FZD5
Entrez Gene ID
7855
Gene Name
frizzled class receptor 5
Gene Aliases
C2orf31, HFZ5
Chromosome Location
Chr. 2: 207762586 - 207769419 on Build GRCh38
UniGene ID
Hs.17631
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003468.3 NP_003459.2 2 2247
GenBank mRNA AB043702.1 - 2 2222
BC114346.2 - 1 1890
U43318.1 - 2 2157

View Full Product Details