Gene Transcripts
- Gene Symbol
- CERS4
- Entrez Gene ID
- 79603
- Gene Name
- ceramide synthase 4
- Gene Aliases
- LASS4, Trh1
- Chromosome Location
- Chr. 19: 8209329 - 8262433 on Build GRCh38
- UniGene ID
- Hs.515111
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_024552.2 |
NP_078828.2
|
7, 8 |
834 |
|
XM_011528290.2 |
XP_011526592.1
|
7, 8 |
870 |
|
XM_011528291.2 |
XP_011526593.1
|
7, 8 |
987 |
|
XM_011528292.2 |
XP_011526594.1
|
8, 9 |
1810 |
|
XM_011528293.2 |
XP_011526595.1
|
8, 9 |
986 |
|
XM_011528294.2 |
XP_011526596.1
|
8, 9 |
954 |
|
XM_011528295.2 |
XP_011526597.1
|
6, 7 |
640 |
|
XM_017027303.1 |
XP_016882792.1
|
8, 9 |
1842 |
|
XM_017027304.1 |
XP_016882793.1
|
9, 10 |
1065 |
|
XM_017027305.1 |
XP_016882794.1
|
6, 7 |
701 |
|
GenBank mRNA |
AK022151.1 |
- |
7, 8 |
794 |
|
AK055414.1 |
- |
4, 5 |
1371 |
|
AK222621.1 |
- |
7, 8 |
801 |
|
AK225394.1 |
- |
7, 8 |
797 |
|
BC009828.2 |
- |
6, 7 |
637 |
3936998 |
View Full Product Details