Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009409.2 | NP_033435.2 | 2 | 547 | |
GenBank mRNA | AK041054.1 | - | 2 | 574 | |
AK087535.1 | - | 2 | 576 | ||
AK163043.1 | - | 2 | 576 | ||
BC041106.1 | - | 2 | 567 | 5368508 | |
BC054541.1 | - | 2 | 547 | 6406462 | |
D38046.1 | - | 2 | 120 |