Gene Transcripts

Gene Symbol
Rhoa
Entrez Gene ID
11848
Gene Name
ras homolog gene family, member A
Gene Aliases
Arha, Arha1, Arha2
Chromosome Location
Chr. 9: 108306205 - 108337939 on Build GRCm38
UniGene ID
Mm.318359
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016802.4 NP_058082.2 6 2116
GenBank mRNA AK083624.1 - 6 1923

View Full Product Details