Gene Transcripts
- Gene Symbol
- Ppp1r2
- Entrez Gene ID
- 66849
- Gene Name
- protein phosphatase 1, regulatory (inhibitor) subunit 2
- Gene Aliases
- 0610025N14Rik, 2310007G06Rik, 4930440J04Rik, 5430408E15Rik, D16Ertd248e, IPP-2
- Chromosome Location
- Chr. 16: 31251541 - 31275277 on Build GRCm38
- UniGene ID
- Mm.291593
- Species
- Mus musculus
- Gene Symbol
- Ppp1r2-ps3
- Entrez Gene ID
- 546723
- Gene Name
- protein phosphatase 1, regulatory (inhibitor) subunit 2, pseudogene 3
- Gene Aliases
- EG546723, Gm5972
- Chromosome Location
- Chr. 19: 30538876 - 30539679 on Build GRCm38
- UniGene ID
- Mm.301509
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC100445.1 |
- |
1 |
407 |
|
View Full Product Details