Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_029934.3 | NP_084210.2 | 8 | 2728 | |
GenBank mRNA | AB055410.1 | - | 8 | 2726 | |
AK150010.1 | - | 8 | 2122 | ||
AK154606.1 | - | 8 | 2727 | ||
AK158998.1 | - | 6 | 3649 | ||
AK170124.1 | - | 8 | 2205 | ||
AK211361.1 | - | 1 | 235 | ||
BC023417.1 | - | 8 | 2176 | 5028759 |