Gene Transcripts
- Gene Symbol
- Kdm5a
- Entrez Gene ID
- 214899
- Gene Name
- lysine (K)-specific demethylase 5A
- Gene Aliases
- AA409370, C76986, Jarid1a, RBP2, Rbbp2
- Chromosome Location
- Chr. 6: 120364099 - 120444574 on Build GRCm38
- UniGene ID
- Mm.404761
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_145997.2 |
NP_666109.2
|
2, 3 |
766 |
|
GenBank mRNA |
AK041568.1 |
- |
2, 3 |
766 |
|
AK046187.1 |
- |
2, 3 |
716 |
|
AK133589.1 |
- |
2, 3 |
435 |
|
AK139619.1 |
- |
2, 3 |
733 |
|
AK144877.1 |
- |
2, 3 |
737 |
|
AK154025.1 |
- |
2, 3 |
422 |
|
AK159079.1 |
- |
2, 3 |
743 |
|
AK166055.1 |
- |
2, 3 |
747 |
|
BC010717.1 |
- |
2, 3 |
407 |
|
BC011506.3 |
- |
2, 3 |
700 |
|
BC080691.1 |
- |
2, 3 |
452 |
|
View Full Product Details