Gene Transcripts

Gene Symbol
BC049762
Entrez Gene ID
193286
Gene Name
cDNA sequence BC049762
Gene Aliases
4930503F14
Chromosome Location
Chr. 11: 51253645 - 51265635 on Build GRCm38
UniGene ID
Mm.80975
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177567.3 NP_808235.1 3 773
GenBank mRNA AK076829.1 - 3 774
AK154967.1 - 4 957
BC049762.1 - 3 650 6774041

View Full Product Details