Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 153864742-153865243 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
L1CAM
Entrez Gene ID
3897
Gene Name
L1 cell adhesion molecule
Gene Aliases
CAML1, CD171, HSAS, HSAS1, MASA, MIC5, N-CAM-L1, N-CAML1, NCAM-L1, S10, SPG1
UniGene
Hs.522818
Gene Chromosome Location
Chr.X: 153861514-153886174 on Build GRCh38
Transcripts
NM_000425.4 NM_001143963.2 NM_001278116.1 NM_024003.3

SNP Detail

SNP ID Chromosome Location
rs782303385 Chr.X: 153865039-153865039 on Build GRCh38
rs148187017 Chr.X: 153865107-153865107 on Build GRCh38
rs375705679 Chr.X: 153864948-153864948 on Build GRCh38
rs950213549 Chr.X: 153865085-153865085 on Build GRCh38
rs201986895 Chr.X: 153865044-153865044 on Build GRCh38
rs200465059 Chr.X: 153864991-153864991 on Build GRCh38
rs886044872 Chr.X: 153865111-153865111 on Build GRCh38
rs782575213 Chr.X: 153865065-153865065 on Build GRCh38
rs782032678 Chr.X: 153864925-153864925 on Build GRCh38
rs782781265 Chr.X: 153864961-153864961 on Build GRCh38
rs781875773 Chr.X: 153864990-153864990 on Build GRCh38
rs782499647 Chr.X: 153864892-153864892 on Build GRCh38
rs374991970 Chr.X: 153865122-153865122 on Build GRCh38
rs782808218 Chr.X: 153865216-153865216 on Build GRCh38
rs398123365 Chr.X: 153864853-153864853 on Build GRCh38
rs559818808 Chr.X: 153865135-153865135 on Build GRCh38
rs201128366 Chr.X: 153864851-153864851 on Build GRCh38
rs888187370 Chr.X: 153864854-153864854 on Build GRCh38
rs781979227 Chr.X: 153864905-153864905 on Build GRCh38
rs879253717 Chr.X: 153864893-153864893 on Build GRCh38
rs782566102 Chr.X: 153865144-153865144 on Build GRCh38
rs372925986 Chr.X: 153864904-153864904 on Build GRCh38
rs782391071 Chr.X: 153865129-153865129 on Build GRCh38
rs200184416 Chr.X: 153865057-153865057 on Build GRCh38
rs782817224 Chr.X: 153865043-153865043 on Build GRCh38
rs201237768 Chr.X: 153865061-153865061 on Build GRCh38
rs782430495 Chr.X: 153864898-153864898 on Build GRCh38
rs782773263 Chr.X: 153864794-153864794 on Build GRCh38
rs782315098 Chr.X: 153865142-153865142 on Build GRCh38
rs782143967 Chr.X: 153864847-153864847 on Build GRCh38
rs782396488 Chr.X: 153864924-153864924 on Build GRCh38
rs782089821 Chr.X: 153864791-153864791 on Build GRCh38
rs201208121 Chr.X: 153865022-153865022 on Build GRCh38
rs782249387 Chr.X: 153865128-153865128 on Build GRCh38
rs398123364 Chr.X: 153864988-153864988 on Build GRCh38
rs372416154 Chr.X: 153865077-153865077 on Build GRCh38
rs998523939 Chr.X: 153864938-153864938 on Build GRCh38
rs201106469 Chr.X: 153864883-153864883 on Build GRCh38
rs782573655 Chr.X: 153864899-153864899 on Build GRCh38
rs782617996 Chr.X: 153865125-153865125 on Build GRCh38
rs782279903 Chr.X: 153864897-153864897 on Build GRCh38
rs398123363 Chr.X: 153865079-153865079 on Build GRCh38
rs376575767 Chr.X: 153864784-153864784 on Build GRCh38
rs782135388 Chr.X: 153865051-153865051 on Build GRCh38
rs782089051 Chr.X: 153865150-153865150 on Build GRCh38
rs782606921 Chr.X: 153865002-153865002 on Build GRCh38
rs967784115 Chr.X: 153865027-153865027 on Build GRCh38
rs782319349 Chr.X: 153864783-153864783 on Build GRCh38
rs782241896 Chr.X: 153865009-153865009 on Build GRCh38
rs782712766 Chr.X: 153864947-153864947 on Build GRCh38
rs202235510 Chr.X: 153864891-153864891 on Build GRCh38
rs782052165 Chr.X: 153865074-153865074 on Build GRCh38
rs782294327 Chr.X: 153865140-153865140 on Build GRCh38
rs782808723 Chr.X: 153864978-153864978 on Build GRCh38
rs149420127 Chr.X: 153864867-153864867 on Build GRCh38
rs1057518490 Chr.X: 153864956-153864956 on Build GRCh38
rs200863731 Chr.X: 153864879-153864879 on Build GRCh38
rs926069186 Chr.X: 153865038-153865038 on Build GRCh38
rs141213959 Chr.X: 153865151-153865151 on Build GRCh38
rs1029546977 Chr.X: 153864987-153864987 on Build GRCh38
rs781861416 Chr.X: 153864845-153864845 on Build GRCh38
rs781869593 Chr.X: 153864975-153864975 on Build GRCh38
rs782104287 Chr.X: 153864955-153864955 on Build GRCh38
rs782450184 Chr.X: 153864864-153864864 on Build GRCh38
rs202082978 Chr.X: 153864852-153864852 on Build GRCh38
rs781813303 Chr.X: 153865197-153865197 on Build GRCh38
rs782142637 Chr.X: 153865200-153865200 on Build GRCh38
rs782558335 Chr.X: 153865105-153865105 on Build GRCh38
rs35902890 Chr.X: 153865088-153865088 on Build GRCh38
rs782505404 Chr.X: 153865011-153865011 on Build GRCh38
rs782661249 Chr.X: 153864894-153864894 on Build GRCh38
rs782736446 Chr.X: 153865076-153865076 on Build GRCh38
rs782051017 Chr.X: 153864936-153864936 on Build GRCh38
rs782283987 Chr.X: 153865037-153865037 on Build GRCh38
rs782190446 Chr.X: 153865116-153865116 on Build GRCh38

View Full Product Details