Gene Transcripts

Gene Symbol
Cyp2ab1
Entrez Gene ID
224044
Gene Name
cytochrome P450, family 2, subfamily ab, polypeptide 1
Gene Aliases
EG224044, F730023N20
Chromosome Location
Chr. 16: 20308387 - 20325412 on Build GRCm38
UniGene ID
Mm.276400
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183158.3 NP_898981.2 9 1254
GenBank mRNA AK089408.1 - 9 1257
BC138088.1 - 5 773 8861107
BC145141.1 - 4 631

View Full Product Details