Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_010637.3 | NP_034767.2 | 3 | 1218 | |
GenBank mRNA | AK141244.1 | - | 1 | 130 | |
AK144942.1 | - | 3 | 1216 | ||
AK155343.1 | - | 3 | 1216 | ||
BC010301.1 | - | 3 | 1080 | 3156339 | |
JF277566.1 | - | 2 | 614 | ||
U20344.1 | - | 3 | 924 | ||
U70662.1 | - | 3 | 1212 |