Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001146347.1 | NP_001139819.1 | 2 | 527 | |
NM_026884.1 | NP_081160.1 | 6 | 640 | ||
NM_029978.1 | NP_084254.1 | 3 | 1079 | ||
GenBank mRNA | AK005271.1 | - | 3 | 391 | |
AK131642.1 | - | 3 | 1078 | ||
BC055444.1 | - | 2 | 528 | 4502433 | |
BC093505.1 | - | 665 | 4503230 |