Gene Transcripts

Gene Symbol
LOC102548927
Entrez Gene ID
102548927
Gene Name
high mobility group protein B4-like
Gene Aliases
Chromosome Location
Chr. X: 53234042 - 53235376 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008773285.1 XP_008771507.1 1 387

View Full Product Details