Gene Transcripts

Gene Symbol
Wdr81
Entrez Gene ID
303312
Gene Name
WD repeat domain 81
Gene Aliases
RGD1311334
Chromosome Location
Chr. 10: 62273817 - 62287207 on Build Rnor_6.0
UniGene ID
Rn.163168
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134360.1 NP_001127832.1 1 3628

View Full Product Details