Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_000425.4 | NP_000416.1 |
NM_001143963.2 | NP_001137435.1 | |
NM_001278116.1 | NP_001265045.1 | |
NM_024003.3 | NP_076493.1 | |
GenBank mRNA | AB102653.1 | - |
AI361399.1 | - | |
AK289754.1 | - | |
AY927629.1 | - | |
BC025843.1 | - | |
BC126229.1 | - | |
BC136447.1 | - | |
DA122729.1 | - | |
EF506611.1 | - | |
ENST00000370060.0 | - | |
HY119527.1 | - | |
M55271.1 | - | |
M74387.1 | - | |
M77640.1 | - | |
X58776.1 | - | |
X59847.1 | - |