Gene Transcripts

Gene Symbol
L1CAM
Entrez Gene ID
3897
Gene Name
L1 cell adhesion molecule
Gene Aliases
CAML1, CD171, HSAS, HSAS1, MASA, MIC5, N-CAM-L1, N-CAML1, NCAM-L1, S10, SPG1
Chromosome Location
Chr.X: 153861514 - 153886174 on Build GRCh38
Uni Gene
Hs.522818
Interrogated Sequence Translated Protein
RefSeq NM_000425.4 NP_000416.1
NM_001143963.2 NP_001137435.1
NM_001278116.1 NP_001265045.1
NM_024003.3 NP_076493.1
GenBank mRNA AB102653.1 -
AI361399.1 -
AK289754.1 -
AY927629.1 -
BC025843.1 -
BC126229.1 -
BC136447.1 -
DA122729.1 -
EF506611.1 -
ENST00000370060.0 -
HY119527.1 -
M55271.1 -
M74387.1 -
M77640.1 -
X58776.1 -
X59847.1 -

View Full Product Details