Gene Transcripts

Gene Symbol
L1cam
Entrez Gene ID
16728
Gene Name
L1 cell adhesion molecule
Gene Aliases
CD171, L1, N-CAM-L1, NCAM-L1
Chromosome Location
Chr.X: 73853780 - 73880841 on Build GRCm38
Uni Gene
Mm.260568
Interrogated Sequence Translated Protein
RefSeq NM_008478.3 NP_032504.3
GenBank mRNA AJ627046.1 -
AK139407.1 -
AK212283.1 -
BC056988.1 -
CJ109973.1 -
CJ258952.1 -
X12875.1 -

View Full Product Details