Gene Transcripts
- Gene Symbol
- LPCAT4
- Entrez Gene ID
- 254531
- Gene Name
- lysophosphatidylcholine acyltransferase 4
- Gene Aliases
- AGPAT7, AYTL3, LPAAT-eta, LPEAT2
- Chromosome Location
- Chr. 15: 34358888 - 34367196 on Build GRCh38
- UniGene ID
- Hs.352614
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK001119.1 |
- |
1 |
460 |
|
View Full Product Details