Gene Transcripts

Gene Symbol
Gins2
Entrez Gene ID
272551
Gene Name
GINS complex subunit 2 (Psf2 homolog)
Gene Aliases
2210013I18Rik, 4833427B12Rik, AI323585, Pfs2
Chromosome Location
Chr. 8: 120581266 - 120589075 on Build GRCm38
UniGene ID
Mm.275910
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178856.1 NP_849187.1 1 109
GenBank mRNA AK014776.1 - 1 123
AK028375.1 - 1 61
AK030758.1 - 126
BC082565.1 - 1 109 30551463

View Full Product Details