Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004581.5 | NP_004572.3 | 8 | 1345 | |
NM_182836.2 | NP_878256.1 | 9 | 1028 | ||
GenBank mRNA | AK291347.1 | - | 9 | 912 | |
AK292613.1 | - | 8 | 1077 | ||
AK303247.1 | - | 7 | 790 | ||
AK308715.1 | - | 9 | 911 | ||
BC003093.1 | - | 9 | 966 | 3537388 | |
CR541797.1 | - | 7 | 833 | ||
Y08200.1 | - | 8 | 1107 |