Gene Transcripts
- Gene Symbol
- Gm7467
- Entrez Gene ID
- 665055
- Gene Name
- predicted gene 7467
- Gene Aliases
- EG665055
- Chromosome Location
- Chr. 5: 76304882 - 76305532 on Build GRCm38
- UniGene ID
- Mm.425276
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK142576.1 |
- |
1 |
606 |
|
View Full Product Details