Gene Transcripts

Gene Symbol
Ppm1j
Entrez Gene ID
295341
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent, 1J
Gene Aliases
Chromosome Location
Chr. 2: 207262886 - 207268601 on Build Rnor_6.0
UniGene ID
Rn.12833
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005540.1 NP_001005540.1 3 555
XM_008761380.1 XP_008759602.1 4 1053
XM_008761381.1 XP_008759603.1 4 1053
GenBank mRNA BC082053.1 - 3 555

View Full Product Details