Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001301645.1 | NP_001288574.1 | 2 | 252 | |
NM_004744.4 | NP_004735.2 | 2 | 446 | ||
GenBank mRNA | AF071510.1 | - | 2 | 285 | |
AK292598.1 | - | 2 | 446 | ||
AY546085.1 | - | 1 | 520 | ||
AY546086.1 | - | 2 | 417 | ||
BC031053.1 | - | 2 | 255 | 5272486 |