Gene Transcripts
- Gene Symbol
- ME3
- Entrez Gene ID
- 10873
- Gene Name
- malic enzyme 3
- Gene Aliases
- NADP-ME
- Chromosome Location
- Chr. 11: 86441108 - 86674745 on Build GRCh38
- UniGene ID
- Hs.199743
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001014811.1 |
NP_001014811.1
|
6, 7 |
1067 |
|
NM_001161586.1 |
NP_001155058.1
|
7, 8 |
1140 |
|
NM_006680.2 |
NP_006671.2
|
7, 8 |
915 |
|
XM_005273717.1 |
XP_005273774.1
|
7, 8 |
1016 |
|
XM_017017136.1 |
XP_016872625.1
|
7, 8 |
1898 |
|
XM_017017137.1 |
XP_016872626.1
|
3, 4 |
440 |
|
XM_017017138.1 |
XP_016872627.1
|
8, 9 |
1024 |
|
GenBank mRNA |
AA403046.1 |
- |
2, 3 |
155 |
|
AA449933.1 |
- |
2, 3 |
155 |
|
AK092181.1 |
- |
7, 8 |
878 |
|
AK300974.1 |
- |
7, 8 |
1140 |
|
AK313695.1 |
- |
7, 8 |
990 |
|
AY424278.1 |
- |
5, 6 |
627 |
|
BC022472.2 |
- |
6, 7 |
1067 |
4812045 |
X79440.1 |
- |
6, 7 |
845 |
|
View Full Product Details