Gene Transcripts

Gene Symbol
SCP2
Entrez Gene ID
6342
Gene Name
sterol carrier protein 2
Gene Aliases
NLTP, NSL-TP, SCP-2, SCP-CHI, SCP-X, SCPX
Chromosome Location
Chr. 1: 52927229 - 53051617 on Build GRCh38
UniGene ID
Hs.476365
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007099.2 NP_001007100.1 2 162
NM_001007100.2 NP_001007101.1 2 153
NM_001007250.2 NP_001007251.1 2 162
NM_001193599.1 NP_001180528.1 12 1387
NM_001193600.1 NP_001180529.1 12 1327
NM_001193617.1 NP_001180546.1 12 1401
NM_002979.4 NP_002970.2 13 1459
XM_005271103.4 XP_005271160.1 13 1402
XM_017002046.1 XP_016857535.1 3 727
GenBank mRNA AB208789.1 - 14 4154
AK294631.1 - 12 1330
AK295214.1 - 12 1344
AK303400.1 - 2 123
AK312856.1 - 13 1410
BC005911.1 - 2 152
BX537618.1 - 2 127
BX537619.1 - 2 127
CA867670.1 - 2 119
DB459183.1 - 2 131
M55421.1 - 2 162
M75883.1 - 13 1312
M75884.1 - 5 598
S52450.1 - 2 162

View Full Product Details