Gene Transcripts

Gene Symbol
TSPYL2
Entrez Gene ID
64061
Gene Name
TSPY like 2
Gene Aliases
CDA1, CINAP, CTCL, DENTT, HRIHFB2216, NP79, SE204, TSPX
Chromosome Location
Chr. X: 53082327 - 53088546 on Build GRCh38
UniGene ID
Hs.136164
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022117.3 NP_071400.1 5 1318
XM_006724592.3 XP_006724655.1 5 1351
XM_017029726.1 XP_016885215.1 4 1483
XM_017029727.1 XP_016885216.1 4 1483
GenBank mRNA AB015345.1 - 5 558
AB209182.1 - 2 901
AF254794.1 - 5 1057
AF273046.1 - 5 1308
AK131220.1 - 4 1456
AK292462.1 - 5 1311
AL713652.1 - 5 1297
AY040871.1 - 5 1307
BC001566.1 - 2 115
BC024270.1 - 5 1298 2819684

View Full Product Details