Gene Transcripts

Gene Symbol
CERS4
Entrez Gene ID
79603
Gene Name
ceramide synthase 4
Gene Aliases
LASS4, Trh1
Chromosome Location
Chr. 19: 8209329 - 8262433 on Build GRCh38
UniGene ID
Hs.515111
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK022151.1 - 9 989
AK225394.1 - 9 992
BC009828.2 - 8 832 3936998

View Full Product Details