Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000116.4 | NP_000107.1 | 11 | 1848 | |
NM_001303465.1 | NP_001290394.1 | 10 | 1860 | ||
NM_181311.3 | NP_851828.1 | 10 | 1758 | ||
NM_181312.3 | NP_851829.1 | 10 | 1806 | ||
NM_181313.3 | NP_851830.1 | 9 | 1716 | ||
NR_024048.2 | 10 | 1864 | |||
XM_006724836.1 | XP_006724899.1 | 11 | 1879 | ||
XM_006724837.1 | XP_006724900.1 | 8 | 1864 | ||
XM_006724839.1 | XP_006724902.1 | 9 | 1747 | ||
XM_006724841.3 | XP_006724904.1 | 11 | 1600 | ||
XM_006724842.3 | XP_006724905.1 | 10 | 1510 | ||
XM_011531189.1 | XP_011529491.1 | 8 | 1666 | ||
XM_011531191.2 | XP_011529493.1 | 10 | 2038 | ||
XM_017029761.1 | XP_016885250.1 | 8 | 1810 | ||
XM_017029762.1 | XP_016885251.1 | 10 | 1789 | ||
XM_017029763.1 | XP_016885252.1 | 8 | 1612 | ||
XM_017029764.1 | XP_016885253.1 | 9 | 1377 | ||
XM_017029765.1 | XP_016885254.1 | 9 | 1915 | ||
GenBank mRNA | AK130900.1 | - | 5 | 2211 | |
BC005062.1 | - | 10 | 1821 | ||
BC011515.1 | - | 7 | 1529 | 4339983 | |
X92762.1 | - | 11 | 1810 |