Gene Transcripts
- Gene Symbol
- CERS3
- Entrez Gene ID
- 204219
- Gene Name
- ceramide synthase 3
- Gene Aliases
- ARCI9, LASS3
- Chromosome Location
- Chr. 15: 100400395 - 100544982 on Build GRCh38
- UniGene ID
- Hs.662371
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001290341.2 |
NP_001277270.1
|
6, 7 |
980 |
|
NM_001290342.2 |
NP_001277271.1
|
5, 6 |
806 |
|
NM_001290343.1 |
NP_001277272.1
|
5, 6 |
717 |
|
NM_178842.4 |
NP_849164.2
|
5, 6 |
713 |
|
XM_011521355.2 |
XP_011519657.1
|
6, 7 |
1023 |
|
XM_011521357.2 |
XP_011519659.1
|
6, 7 |
917 |
|
XM_011521358.1 |
XP_011519660.1
|
6, 7 |
801 |
|
XM_017022002.1 |
XP_016877491.1
|
5, 6 |
797 |
|
XM_017022003.1 |
XP_016877492.1
|
5, 6 |
722 |
|
XM_017022004.1 |
XP_016877493.1
|
4, 5 |
633 |
|
GenBank mRNA |
BC028703.1 |
- |
5, 6 |
794 |
|
BC034500.1 |
- |
5, 6 |
672 |
4838423 |
BC034970.1 |
- |
5, 6 |
715 |
4822006 |
View Full Product Details