Gene Transcripts
- Gene Symbol
- Hpn
- Entrez Gene ID
- 15451
- Gene Name
- hepsin
- Gene Aliases
- Hlb320
- Chromosome Location
- Chr. 7: 31098725 - 31115326 on Build GRCm38
- UniGene ID
- Mm.19182
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF030065.1 |
- |
9 |
950 |
|
AY234104.1 |
- |
8 |
1056 |
|
View Full Product Details