Gene Transcripts

Gene Symbol
Krtap6-1
Entrez Gene ID
16700
Gene Name
keratin associated protein 6-1
Gene Aliases
HGTpII.1, HGTpII.2
Chromosome Location
Chr. 16: 89031699 - 89032292 on Build GRCm38
UniGene ID
Mm.6973
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010672.3 NP_034802.1 1 509
GenBank mRNA CJ048388.1 - 1 549
D86420.1 - 1 509

View Full Product Details